Learn More
ANP32B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10843-1-AP-20UL
Beschreibung
Multifunctional protein working as a cell cycle progression factor as well as a cell survival factor. Required for the progression from the G1 to the S phase. Anti-apoptotic protein which functions as a caspase-3 inhibitor. Has no phosphatase 2A (PP2A) inhibitor activity.Spezifikation
ANP32B | |
Polyclonal | |
Unconjugated | |
ANP32B | |
ANP32B, APRIL, PHAPI2, Silver stainable protein SSP29, SSP29 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10541, 67628 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q92688, Q9EST5 | |
ANP32B | |
ANP32B Fusion Protein Ag1274 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.