Learn More
Sampling Systems™ Angular Beaker
These angular beaker attachments allow the cup to be set in specific locations while using the TeleScoop.
Marke: Sampling Systems™ 5065B-252
Beschreibung
- Telescopic samplers are often used for taking surface samples from rivers, tanks and lakes
- A wide choice of attachments is available to make the TeleScoop ideally suited for most applications
- Attachments are interchangeable and simply click into position
Spezifikation
Swabbing Template | |
Autoklavierbar | |
Reusable | |
Unsteril |
Polypropylen | |
250 mL | |
Yes |