Learn More
BINDER™ Analog Output
Non-adjustable analog output
Brand:  BINDER™ 8012-2574
 View more versions of this product
Product Code. 30238280
  • 687.32 CHF / 1 Stück
Verfügbar ab: 07-05-2025
Sie sich ein, um den Lagerbestand zu sehen
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Analog output
  • Not adjustable
  • For temperature values or temperature and humidity values
  • 4 to 20 mA
Specifications
Specifications
KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600
Copper
Analog Output
For Temperature and Humidity Values
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BINDER™ Analog Output > For use with KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600