Learn More
alpha-synuclein Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
136.48 CHF - 385.92 CHF
Spezifikation
Antigen | alpha-synuclein |
---|---|
Konzentration | 0.51 mg/mL |
Anwendungen | Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | ||||||
16897493
|
Proteintech
10842-1-AP-20UL |
20 μL |
136.48 CHF
20 Mikroliter |
Lager vorrätig
Loggen Sie sich ein, um den Lagerbestand zu sehen |
Sie haben bereits ein Konto? Melden Sie sich an um den Preis und die Verfügbarkeit zu sehen. Neu hier? Registrieren Sie sich noch heute bei uns! |
|||||
16887493
|
Proteintech
10842-1-AP-150UL |
150 μL |
385.92 CHF
150 Mikroliter |
Lager vorrätig
Loggen Sie sich ein, um den Lagerbestand zu sehen |
Sie haben bereits ein Konto? Melden Sie sich an um den Preis und die Verfügbarkeit zu sehen. Neu hier? Registrieren Sie sich noch heute bei uns! |
|||||
Beschreibung
This antibody can recognize all the isoforms of alpha-syn.
Alpha-Synuclein belongs to the Synuclein family, which includes beta and gamma Synuclein, and is predomitly expressed in neurons, and concentrated at synaptic terminals. Synucleins may be involved in the regulation of dopamine release and transport and the modulation of synaptic vesicle function. Alpha-Synuclein is a small, presynaptic protein that is the major nonamyloid component of the pathological inclusions characteristic of a wide range of neurodegenerative disorders, collectively known as synucleinopathies. Alpha-Synuclein is involved in the formation of SNARE complexes, and most significantly, aggregated alpha-synuclein is one of the major components found in the Lewy bodies that occur in Parkinson′s disease (PD) and other neurodegenerative disorders. Mutation of the alpha-synuclein gene is associated with familial forms of PD. Alpha-Synuclein is known to reduce the fibrillization of the microtubule-associated protein, tau, to bind various targets such as 14-3-3, protein kinase C, synphilin-1, Elk and Tat-binding protein 1. The alpha and beta synucleins are found primarily in brain tissue where they are seen mainly in neuronal cytosol and presynaptic terminals. The antibody specific for alpha-synuclein does not cross react with beta-synuclein and will be useful for the confirmation of Parkinson′s disease and Lewy body dementia and other studies for the localization and detection of alpha-synuclein. Four alternatively spliced transcripts that encode two different isoforms of the alpha-synuclein protein have been identified.Spezifikation
alpha-synuclein | |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
O55042, P37377, P37840 | |
20617, 29219, 6622 | |
alpha-synuclein Fusion Protein Ag1285 | |
Primary | |
-20°C | |
SNCA |
0.51 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Alpha synuclein, NACP, PARK1, PARK4, SNCA | |
SNCA | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.