Learn More
alpha-synuclein Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 10842-1-AP-20UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16897493
  • 136.48 CHF / 20 Mikroliter
Lager vorrätig
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
This antibody can recognize all the isoforms of alpha-syn.
Alpha-Synuclein belongs to the Synuclein family, which includes beta and gamma Synuclein, and is predomitly expressed in neurons, and concentrated at synaptic terminals. Synucleins may be involved in the regulation of dopamine release and transport and the modulation of synaptic vesicle function. Alpha-Synuclein is a small, presynaptic protein that is the major nonamyloid component of the pathological inclusions characteristic of a wide range of neurodegenerative disorders, collectively known as synucleinopathies. Alpha-Synuclein is involved in the formation of SNARE complexes, and most significantly, aggregated alpha-synuclein is one of the major components found in the Lewy bodies that occur in Parkinson′s disease (PD) and other neurodegenerative disorders. Mutation of the alpha-synuclein gene is associated with familial forms of PD. Alpha-Synuclein is known to reduce the fibrillization of the microtubule-associated protein, tau, to bind various targets such as 14-3-3, protein kinase C, synphilin-1, Elk and Tat-binding protein 1. The alpha and beta synucleins are found primarily in brain tissue where they are seen mainly in neuronal cytosol and presynaptic terminals. The antibody specific for alpha-synuclein does not cross react with beta-synuclein and will be useful for the confirmation of Parkinson′s disease and Lewy body dementia and other studies for the localization and detection of alpha-synuclein. Four alternatively spliced transcripts that encode two different isoforms of the alpha-synuclein protein have been identified.
Spezifikation
Spezifikation
alpha-synuclein
Polyclonal
Unconjugated
SNCA
Alpha synuclein, NACP, PARK1, PARK4, SNCA
Rabbit
Antigen Affinity Chromatography
RUO
20617, 29219, 6622
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin)
0.51 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O55042, P37377, P37840
SNCA
alpha-synuclein Fusion Protein Ag1285
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote
Berichtigung von Produktinhalten
Name des Produkts
alpha-synuclein Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated