Learn More
Tocris Bioscience™ Xanomeline oxalate
Functionally selective M1 agonist
Marke: Tocris Bioscience™ 3569/10
222.22 CHF gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Xanomeline oxalate is a functionally selective muscarinic M1 receptor agonist (EC50 values are 0.3, 5, 42, 52 and 92.5 nM at M1, M3, M5, M4 and M2 receptors respectively). Displays a complex pharmacological profile: reversible and wash-resistant binding, resulting in full agonist activity at M1; delayed wash-resistant partial agonist activity at M2; and delayed wash-resistant full agonist activity at M4. Exhibits antipsychotic activity, and improves cognitive deficits and behavioral disturbances in Alzheimer's disease and schizophrenia.Spezifikation
>98% | |
Xanomeline oxalate | |
Xanomeline oxalate |
10 mg | |
371.46 |
Nur für Forschungszwecke