Learn More
Cofra Women's Safety Shoes, Paula Series
Women's Safety Shoes have breathable leather uppers for comfort, steel toecaps for extra protection and tough anti-static eva-nitrile foodbeds.
Marke: Cofra 76590-000_35
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Lace-up general purpose safety shoes
- Water-repellant breathable black nubuck uppers
- Steel toecaps resistant to 200J
- Tough EVA-nitrile anti-static footbed
- Breathable polyamide internal lining
- Slip-resistant polyurethane soles
Spezifikation
Lace-up | |
EVA-Nitrile | |
Leather | |
Anti-slip | |
Stahl | |
Frauen |
Black | |
Polyamide | |
2 | |
Polyurethan | |
EN ISO 20345:2011 | |
Wasser |