missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR40A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20478-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody specifically recognizes the 51 kDa protein.
Cancer-testis (CT) antigens are a type of antigen whose expression is highly restricted, primarily in normal testis, but aberrantly in different tumor types. Their immunogenicity in cancer patients makes them prime targets for immunotherapy of human tumors. One such CT antigen is TCC52, a novel centrosomal protein. TCC52 is a WD repeat-containing protein that interacts with the COP9 signalosome, a macromolecular complex that interacts with cullin-RING E3 ligases and regulates their activity by hydrolyzing cullin-Nedd8 conjugates. The normal biological function of this protein is not known at this time.Spezifikation
WDR40A | |
Polyclonal | |
Unconjugated | |
DCAF12 | |
DCAF12, KIAA1892, TCC52, WD repeat domain 40A, WDR40A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25853 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5T6F0 | |
DCAF12 | |
WDR40A Fusion Protein Ag14349 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
WDR40A Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido