missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ VitroEase™ Buffer Screening Kit and Detergents Artikelnummer.: 30054542

Thermo Scientific™ VitroEase™ Buffer Screening Kit and Detergents

Artikelnummer. p-200019987 Alle ansehen Thermo Scientific Produkte
CTAB, 2 Stück
Click to view available options
Puffer:
CHAPS
CTAB
DM
FOM
OG
Tween™-20
Packungsgröße:
1 Set
2 Stück
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 30054542

Marke: Thermo Scientific™ A65372

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

The Thermo Scientific VitroEase Buffer Screening Kit provides pre-formulated buffers and detergents and a comprehensive screening strategy to identify optimal protein conditions for single particle cryo-EM sample analysis.

The Thermo Scientific VitroEase Buffer Screening Kit provides pre-formulated buffers and detergents and a comprehensive screening strategy to identify optimal protein conditions for single particle cryo-EM sample analysis.

Features of the VitroEase Buffer Screening Kit include:
Ease of use—14 buffers and 6 detergents pre-formulated at 10X concentration ready to be spiked in with your protein sample with minimal sample dilution
Convenient—buffers and detergents provided with color-coded caps that correlate with color-coded screening strategy
Comprehensive—optimized screening strategy encompassing two rounds of screening with 24 grids with trouble-shooting suggestions
Flexible—wide range of common buffers and detergents for cryo-EM sample preparation sufficient for sample optimization of 400 grids
Validated—components and screening strategy validated with single particle cryo-EM analysis

Cryo-EM sample screening evaluates protein behavior in various buffers and additive conditions with the end goal of identifying the optimal conditions in which the protein is stable, randomly oriented, and equally distributed in ice. Kit buffers are provided with pHs from 3.6 to 8.9 pre-mixed with various salts and divalent cations. Detergents are provided at 10X concentrations with an easy-to-use table to convert volume to CMC concentration.

The comprehensive sample screening strategy uses two rounds of screening and 24 grids, taking approximately two days to process (figure below). In the first round, optimal protein concentration, buffer, and pH conditions are determined. In the second round, detergents are further optimized. A condensed strategy is also provided using 8 grids and taking one day to process, which can be used when little sample is available or for use with side entry or semi-automated sample loading systems, like the Tundra Cryo-TEM.

The recommended screening strategies are designed for use with the Thermo Scientific Vitrobot Mk4 and Krios, Glacios, Arctica microscopes with Autoloader systems and EPU multigrid.

TRUSTED_SUSTAINABILITY

Spezifikation

Chemischer Name oder Material Detergent A (refill)
Physikalische Form Flüssig
Konzentration 0.3% (10X CMC)
Menge 2 x 100 μl
Verpackung Polypropylen-Fläschchen
Puffer CTAB
Zur Verwendung mit (Geräte) Vitrobot™ Mark IV System, Krios™, Glacios™oder Talos™ Arctica™ Mikroskope

Nur für Forschungszwecke.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Thermo Scientific™ VitroEase™ Buffer Screening Kit and Detergents >

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt