missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral HIV-1 Gag p24 Antibody, R&D Systems™
Mouse Monoclonal Antibody
253.00 CHF - 565.00 CHF
Spezifikation
Antigen | HIV-1 Gag p24 |
---|---|
Klon | 1042942 |
Verdünnung | Western Blot 1 ug/mL, ELISA |
Anwendungen | Western Blot, ELISA |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18794367
|
R&D Systems
MAB11189-SP |
25 μg |
253.00 CHF
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18783497
|
R&D Systems
MAB11189-100 |
100 μg |
565.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot, ELISASpezifikation
HIV-1 Gag p24 | |
Western Blot 1 ug/mL, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
155030 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
1042942 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Virus | |
Q77YG1_9HIV1 | |
E. coli-derived recombinant viral HIV-1 Gag p24, Pro149-Gly354, Accession # Q77YG1_9HIV1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Viral HIV-1 Gag p24 Antibody, R&D Systems™