Learn More
VACUUBRAND™ VARIO™ Diaphragm Pump, MV 2 VARIO Select
An excellent solution for continuous, oil-free pumping of non-corrosive gases. The MV 2 VARIO Select provides high flow rates, even near ultimate vacuum. The VACUU·SELECT Vacuum Controller with graphical user interface and predefined applications simplifies work.
6390.00 CHF
Spezifikation
Max. Vakuum | 0,3 mbar |
---|---|
Regelung | VACUU·SELECT |
Enthält | Gebrauchsfertige Pumpe (vollständig montiert, inkl. Handbuch) |
Motordrehzahl | 30 bis 2400 min-1 |
Zertifizierung/Konformität | CE-Zertifizierung; II 3/-G IIC T3 x interne Atm. Nur |
Beschreibung
- For continuous, oil-free pumping of non-corrosive gases to a vacuum level as deep as 0.3 mbar
- Extraordinary diaphragm life for minimum operational and servicing costs
- Contamination-free pumping and evacuation
- Powerful pump with excellent ultimate vacuum
- TURBO·MODE for use as backing pump for turbo pumps
- VARIO technolgy provides pinpoint vacuum control by adjusting the motor speed resulting in exceptionally long maintenance intervals
- VACUU·SELECT Controller is an easy-to-use, efficient controller for laboratory applications
- Pre-defined procedures for individual applications
- Easy drag-and-drop editing for creating new procedures
- Touch screen display can be operated even when wearing safety glove
- Choose from 14 languages
Spezifikation
0,3 mbar | |
Gebrauchsfertige Pumpe (vollständig montiert, inkl. Handbuch) | |
CE-Zertifizierung; II 3/-G IIC T3 x interne Atm. Nur | |
MV 2 VARIO Select | |
0,53 kW | |
40 °C | |
3,3 m3/Std. | |
Membranpumpe | |
4 | |
4 | |
200/230 V |
VACUU·SELECT | |
30 bis 2400 min-1 | |
325 x 235 x 245 mm | |
45 dBA | |
19,3 kg | |
10 °C | |
50 bis 60 Hz | |
IP 20 | |
Intelligentes Vakuum mit VARIO™-Steuerung, Vorvakuumerzeugung für Turbo-Molekularpumpen, OEM-Produkte: Nach individuellen Anforderungen | |
1,1 bar |