missing translation for 'onlineSavingsMsg'
Learn More

Tris(pentafluorophenyl)phosphin, 97 %, Thermo Scientific Chemicals

CAS: 1259-35-4 | C18F15P | 532.15 g/mol

56.38 CHF - 157.00 CHF

Chemische Identifikatoren

CAS 1259-35-4
Summenformel C18F15P
Molekulargewicht (g/mol) 532.15
MDL-Nummer MFCD00079654
InChI-Schlüssel FQLSDFNKTNBQLC-UHFFFAOYSA-N
Synonym tris pentafluorophenyl phosphine, phosphine, tris pentafluorophenyl, tris perfluorophenyl phosphine, tris pentafluorophenyl phosphane, tris 2,3,4,5,6-pentafluorophenyl phosphane, pubchem6422, acmc-1bogg, ksc491k1f, tris-pentafluorophenyl-phosphane
PubChem CID 102076
IUPAC-Name tris(2,3,4,5,6-pentafluorophenyl)phosphane
SMILES FC1=C(F)C(F)=C(P(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Quantity & Availability  
11337825
SDS View Documents
Thermo Scientific Alfa Aesar
L02748.03
1 g
56.38 CHF
1 Gramm
Verfügbar ab: 07-07-2025
Add to basket
 
11347825
SDS View Documents
Thermo Scientific Alfa Aesar
L02748.06
5 g
157.00 CHF
5 Gramm
Lager vorrätig
Add to basket
 
Description

Description

Tris(pentafluorophenyl)phosphine is an additive for high voltage batteries. It acts as building blocks, coupling reagents, derivatizing reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Anwendungen
Tris(pentafluorphenyl)phosphin ist ein Additiv für Hochspannungsbatterien. Es dient als Baustein, Kupplungsreagenz, Derivatisierungsreagenz.

Löslichkeit
Unlöslich in Wasser.

Hinweise
In einem kühlen, trockenen, gut belüfteten Bereich lagern, der sich nicht mit inkompatiblen Substanzen in Verbindung bringt. Unter inertem Gas handhaben. Nicht kompatibel mit starken Oxidationsmitteln.
Specifications
Mehr anzeigen
Videos
Safety and Handling

Safety and Handling

GHS H-Satz
H315-H319-H335
Verursacht Hautreizungen.
Verursacht schwere Augenreizung.
Kann die Atemwege reizen.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

Gehäuse H315-H319-H335

missing translation for 'einecsNumber' : 215-021-2

missing translation for 'tsca' : Nein

Recommended Storage : Umgebungstemperaturen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tris(pentafluorophenyl)phosphin, 97 %, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.