missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TIM-3 Antibody Pair [HRP]
Monoclonal Antibody Pair
Marke: Novus Biologicals™ NBP2-79594-5Plates
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TIM-3.Spezifikation
31.25 pg/mL | |
Sandwich ELISA | |
Sandwich ELISA Kit | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TIM-3. | |
Sandwich ELISA | |
84868 | |
Rabbit/Mouse | |
5 Plates | |
Immunology, Innate Immunity | |
TIM-3 |
31.25-2000 pg/mL | |
HRP | |
Antibody Pairs | |
Human | |
CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 | |
HAVCR2 | |
RUO | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 ug/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2000 pg/mL is recommended. |