missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TIM-3 Antibody Pair [HRP]

Monoclonal Antibody Pair

Marke:  Novus Biologicals™ NBP2-79594-15PLATES

 Lernen Sie weitere Varianten dieses

Artikelnummer. 18290878

  • 1265.00 CHF / 15 Stück
Verfügbar ab: 03-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TIM-3.
Spezifikation

Spezifikation

31.25 pg/mL
Sandwich ELISA
Sandwich-ELISA-Kit
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TIM-3.
Sandwich-ELISA
84868
Rabbit/Mouse
15 Plates
Immunology, Innate Immunity
TIM-3
31.25-2000 pg/mL
HRP
Antibody Pairs
Human
CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3
HAVCR2
  • Standard: Each vial contains 85 ng of recombinant Human TIM-3.
  • Capture Antibody: 1 mg/mL of rabbit anti-Human TIM-3 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of mouse anti-Human TIM-3 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • RUO
    Storage is content dependent.
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2000 pg/mL is recommended.
    Produktvorschläge

    Produktvorschläge

    Videos
    Sicherheitsdatenblatt (SDS)
    Dokumentation

    Dokumentation

    Zertifikate
    Sonderangebote

    Sonderangebote