missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TIM-1/KIM-1/HAVCR Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
Marke: Novus Biologicals™ NBP2-79564-15PLATES
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This solid phase sandwich ELISA utilizes an antibody specific for Mouse TIM-1/KIM-1/HAVCR.Spezifikation
18.75 pg/mL | |
Sandwich ELISA | |
Sandwich-ELISA-Kit | |
This solid phase sandwich ELISA utilizes an antibody specific for Mouse TIM-1/KIM-1/HAVCR. | |
Sandwich-ELISA | |
26762 | |
Rabbit | |
15 Plates | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1200 pg/mL is recommended. |
18.75-1200 pg/mL | |
HRP | |
Antibody Pairs | |
Maus | |
HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
HAVCR1 | |
RUO | |
TIM-1/KIM-1/HAVCR |