missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Thermus thermophilus cas3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5117525
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody has been tested in direct-ELISA.
CRISPR (clustered regularly interspaced short palindromic repeat), is an adaptive immune system that provides protection against mobile genetic elements (viruses, transposable elements and conjugative plasmids). CRISPR clusters contain sequences complementary to antecedent mobile elements and target invading nucleic acids. CRISPR clusters are transcribed and processed into CRISPR RNA (crRNA). Cas3 plus Cascade participate in CRISPR interference, the third stage of CRISPR immunity. The N-terminal domain (residues 6-260) acts as a ssDNA endonuclease, has no activity on dsDNA.
Spezifikation
Thermus thermophilus cas3 | |
Polyclonal | |
Unconjugated | |
ygcB | |
CRISPR-associated endonuclease; helicase Cas3 | |
Rabbit | |
Protein G | |
RUO | |
3167883, 947229 | |
-20°C or -80°C if preferred | |
Liquid |
Western Blot | |
4.95 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P38036, Q53VY2 | |
cas3, ygcB | |
Recombinant Thermus thermophilus CRISPR-associated endonuclease/helicase Cas3 protein (120-226aa). | |
100 μL | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Invitrogen™ Thermus thermophilus cas3 Polyclonal Antibody > 100 μL; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur