missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF beta 1/2/3 Antibody (1D11), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals FAB1835N
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TGF beta 1/2/3 Monoclonal antibody specifically detects TGF beta 1/2/3 in Broad Species Reactivity samples. It is validated for Western Blot, Binding Inhibition, ELISA Standard (Matched Pair), Neutralization, ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair)
Spezifikation
TGF beta 1/2/3 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7040 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Binding Inhibition Assay, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair) | |
1D11 | |
Western Blot, Binding Inhibition, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair) | |
CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor, beta 1 | |
Bovine bone-derived TGF-beta 1 and TGF-beta 2 | |
0.1 mL | |
Primary | |
Broad Species Reactivity | |
Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
TGF beta 1/2/3 Antibody (1D11), Alexa Fluor™ 700, Novus Biologicals™ > 0.1 mL, Alexa Fluor 700
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur