Learn More
Kern Sohn Stereo-Zoom-Mikroskop OZM
First-class optics and strong illumination combined with a high level of flexibility.
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
1328.45 CHF - 1653.39 CHF
Spezifikation
Spannung | 230 V |
---|---|
Vergrößerungsleistung | 0.7X to 4.5X |
Beleuchtungsart | LED |
Produktcode | Marke | Kopfausführung | Preis | Menge & Verfügbarkeit | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Kopfausführung | Preis | Menge & Verfügbarkeit | ||||||
17359541
![]() |
Kern Sohn
OZM 542 |
Binokular |
1328.45 CHF
1 Stück |
Verfügbar ab: 21-02-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||||
17369541
![]() |
Kern Sohn
OZM 544 |
Trinokular |
1653.39 CHF
1 Stück |
Verfügbar ab: 21-02-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||||
Beschreibung
- The KERN OZM-5 series is a range of excellent stereo zoom microscopes with above-average optical features.
- The ergonomic shape allows a simple, effortless working over a period of several hours.
- The extraordinarily strong and continuously dimmable 3W LED incident and transmitted illumination ensures a flexible and particularly good level of illumination for your sample.
- With its large working distance, an extra large field of view and its brilliant resolution, the KERN OZM provides sharp, high-contrast, colour-true images.
- The zoom objective offers you continuous magnification from 7×–45×.
- The pillar stand is particularly flexible due to its variable and sturdy adjustment mechanism and therefore enables ergonomic working procedures.
- A large selection of eyepieces, (universal) stands, a darkfield kit, external illumination units as well as auxiliary objectives and more are available as accessories
- A protective dust cover, eye cups, as well as multi-lingual user instructions are included in the scope of delivery.
- Scope of application: in vitro fertilisation, detection of parasites, zoology and botany, tissue preparation, section, quality control, Electronics and semiconductor industry, assembly and repair
- Applications/Samples: Samples with focus on three-dimensional impression, zoom with variable magnification (depth, thickness), e.g. insects, seeds, circuit boards, components
Spezifikation
230 V | |
0.7X to 4.5X | |
LED |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Das Fisher Scientific Encompass-Programm bietet Artikel an, die nicht Teil unseres Vertriebsportfolios sind. Diese Produkte verfügen in der Regel nicht über Bilder oder detaillierte Beschreibungen. Wir sind jedoch bestrebt, Ihr Einkaufserlebnis zu verbessern. Bitte verwenden Sie das untenstehende Formular, um uns Feedback zum Inhalt dieses Produkts zu geben.
Name des Produkts
Kern Sohn Stereo-Zoom-Mikroskop OZM