missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT6 (Solitary Fibrous Tumor Marker) Mouse, Clone: STAT6/2410, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 6778-MSM1-P0
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
STAT6 (Solitary Fibrous Tumor Marker) | |
Cytoplasmic and Nuclear | |
STAT6/2410 | |
Unconjugated | |
Mouse | |
119kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
6778 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human STAT6 protein | |
HeLa or HepG2 cells. Kidney | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |