Learn More
Cole-Parmer Sanitary Clamp Fittings, Straight Threaded Adapter, 316 Stainless Steel
Great for food, beverage, and dairy applications—fittings comply with FDA and 3A standards
Marke:  Cole-Parmer 50118-22
 Lernen Sie weitere Varianten dieses
Artikelnummer. 30053508
  • 66.55 CHF / 1 Stück
Verfügbar ab: 04-08-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Ideal for general-purpose, food, beverage, and pharmaceutical applications. Free technical application support available to assist with tubing selection. 316L stainless steel (SS) offers excellent chemical resistance against corrosive acids and bases. Connet to pipes using the NPT(M) thread connection
Spezifikation
Spezifikation
3 A;FDA
1-1/2 in. NPT(M)
1 in. Sanitary Clamp Size
Sanitary x Threaded NPT(M)
Straight