Learn More
Thermo Scientific™ Remel™ PathoDX™ RSV Control Slide
Remel PathoDx and RSV Control Slide is used with RSV immunofluorescent assay.
Marke: Thermo Scientific™ R62375
Beschreibung
Remel PathoDx and RSV Control Slide is used with RSV immunofluorescent assay.
Ordering Alerts:
Each slide has 1 positive and 1 negative well.
Spezifikation
PathoDx™ RSV Control Slides | |
Control | |
5 Objektträger/Pk. |