missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Sheep GHRH Protein
A DNA sequence encoding the Ovis aries (Sheep) GHRH, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6084-ec-200ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
GHRH Protein | |
Research Use Only | |
GHRH | |
Recombinant Protein | |
E. coli | |
YADAIFTNSY RKILGQLSAR KLLQDIMNRQ QGERNQEQGA KVRL | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Full Length | |
Sheep | |
GST | |
Tris-based buffer, 50% glycerol |