missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant S. cerevisiae PPA1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marke: R&D Systems™ 8088-PP-050
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Measured by its ability to hydrolyze pyrophosphate. The specific activity is >125,000 pmol/min/μg, as measured under the described conditions.Bestellinformation
The product is shipped with polar packs. Upon receipt, store it immediately at the recommended storage temperatures.

Spezifikation
P00817 | |
Supplied as a 0.2 μm filtered solution in Tris, NaCl, DTT and MgCl. | |
M.W. (Observed): 36 kDa, reducing conditions; M.W. (theoretical): 33 kDa | |
E. coli-derived yeast Inorganic Pyrophosphatase/PPA1 protein Thr2-Val287, with an N-terminal Met and 6-His tag | |
<0.01 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
Inorganic Pyrophosphatase/PPA1 |
Enzyme Activity | |
5464 | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
IPP1 | |
>80%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane) |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
R&D Systems™ Recombinant S. cerevisiae PPA1 Protein > 50ug
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur