missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Integrin alpha 2 beta 1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marke: R&D Systems™ 7828-A2-050
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Recombinant Mouse Integrin alpha 2 beta 1 Protein is derived from CHO. The Recombinant Mouse Integrin alpha 2 beta 1 Protein has been validated for the following applications: Bioactivity.
Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS. | |
130 kDa (Integrin alpha2) & 86.5 kDa (Integrin beta1) | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
16401 | |
Integrin alpha 2 beta 1 | |
Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha 2 beta 1 protein, (N-terminus) Mouse Integrin alpha 2 (Tyr27-Thr1129) Accession &Num; Q62469.2 His-Pro GGGSGGGS Acidic Tail HHHHHH Mouse Integrin beta 1 (Gln21-Asp728) Accession &Num; P09055 His-Pro GGGSGGGS Basic Tail (C-terminus) | |
Noncovalently-linked heterodimer | |
Recombinant |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
R&D Systems™ Recombinant Mouse Integrin alpha 2 beta 1 Protein > 50ug; Carrier Free
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur