Learn More
R&D Systems™ Recombinant Mouse IL-7 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke:  R&D Systems™ 407-ML-200
1443.59 CHF gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
 Lernen Sie weitere Varianten dieses
Artikelnummer. 18793108
  • 1845.00 CHF / 200 Mikrogramm
Verfügbar ab: 10-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The Recombinant Mouse IL-7 Protein is derived from E. coli. The Recombinant Mouse IL-7 Protein has been validated for the following applications: Bioactivity.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
15 kDa
200 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
16196
IL-7
E. coli-derived mouse IL-7 protein Glu26-Ile154,with an N-terminal Met
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.