Learn More
R&D Systems™ Recombinant Mouse Erythropoietin/EPO Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke:  R&D Systems™ 959-ME-010
Artikelnummer. 18314947
  • 549.00 CHF / 10 Mikrogramm
Verfügbar ab: 11-03-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The Recombinant Mouse Erythropoietin Protein is derived from NS0. The Recombinant Mouse Erythropoietin Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
18.6 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
13856
Erythropoietin/EPO
Mouse myeloma cell line, NS0-derived mouse Erythropoietin/EPO protein Ala27-Arg192
ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Recombinant
Produktvorschläge
Dokumentation
One moment while we fetch your results.
Name des Produkts
R&D Systems™ Recombinant Mouse Erythropoietin/EPO Protein > 10ug