Learn More
R&D Systems™ Recombinant Mouse Dkk-4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke:  R&D Systems™ 3105-DK-010
 Lernen Sie weitere Varianten dieses
Artikelnummer. 18156250
  • 569.00 CHF / 10 Mikrogramm
Verfügbar ab: 19-02-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
The Recombinant Mouse Dkk-4 Protein is derived from NS0. The Recombinant Mouse Dkk-4 Protein has been validated for the following applications: Bioactivity.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
23 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
234130
Dkk-4
Mouse myeloma cell line,NS0-derived mouse Dkk-4 protein Leu19-Ile221 (Arg67Ser and Arg70Ser),with a C-terminal 6-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.