missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Serum Response Factor SRF GST (N-Term) Protein
Recombinant Protein
Marke: Novus Biologicals™ H00006722-Q01-10ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Spezifikation
Western Blot, ELISA, SDS-PAGE, Protein Array, Immunoaffinity Purification | |
6722 | |
10 μg | |
MCM1, serum response factor, serum response factor (c-fos serum response element-binding transcriptionfactor) | |
Phospho Specific | |
Serum Response Factor SRF |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
MolecularWeight-theroretical: 37.07 kDa | |
Store at -80°C. Avoid freeze-thaw cycles. | |
SRF | |
>80% by SDS-PAGE and Coomassie blue staining |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Novus Biologicals™ Recombinant Human Serum Response Factor SRF GST (N-Term) Protein > 10 μg
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur