missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLA2G2E Protein
A cDNA sequence encoding the PLA2G2E was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 5645.00 CHF
Spezifikation
Name | PLA2G2E Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15992419
|
enQuireBio™
QP13073-2ug |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 11-12-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15972419
|
enQuireBio™
QP13073-10ug |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 11-12-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15982419
|
enQuireBio™
QP13073-1mg |
1 mg |
5645.00 CHF
1 Milligramm |
Verfügbar ab: 11-12-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
PLA2G2E Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Sterile filtered and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH 4. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKSPHVLVFL CLLVALVTGN LVQFGVMIEK MTGKSALQYN DYGCYCGIGG SHWPVDQTDW CCHAHDCCYG RLEKLGCEPK LEKYLFSVSE RGIFCAGRTT CQRLTCECDK RAALCFRRNL GTYNRKYAHY PNKLCTGPTP PC | |
Greater than 95% as determined by SDS PAGE. |