missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLA2G2E Protein
A cDNA sequence encoding the PLA2G2E was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13073-1mg
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
PLA2G2E Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKSPHVLVFL CLLVALVTGN LVQFGVMIEK MTGKSALQYN DYGCYCGIGG SHWPVDQTDW CCHAHDCCYG RLEKLGCEPK LEKYLFSVSE RGIFCAGRTT CQRLTCECDK RAALCFRRNL GTYNRKYAHY PNKLCTGPTP PC | |
Greater than 95% as determined by SDS PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Sterile filtered and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH 4. |