missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDK1 Protein
A cDNA sequence encoding the PDK1 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 2605.00 CHF
Spezifikation
Name | PDK1 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15972189
|
enQuireBio™
QP12998-10ug |
10 μg |
131.00 CHF
10 Mikrogramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15992189
|
enQuireBio™
QP12998-50ug |
50 μg |
203.00 CHF
50 Mikrogramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15982189
|
enQuireBio™
QP12998-1mg |
1 mg |
2605.00 CHF
1 Milligramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
PDK1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PDK1 solution containing 20mM Tris pH 7, 0.5mM DTT, 0.1M NaCl, 0.1mM EDTA 0.1mM PMSF, 1mM MgCl2 and 40% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSSDSGSSPA SERGVPGQVD FYARFSPSPL SMKQFLDFGS VNACEKTSFM FLRQELPVRL ANIMKEISLLPDNLLRTPSV QLVQSWYIQS LQELLDFKDK SAEDAKAIYD FTDTVIRIRN RHNDVIPTMA QGVIEYKESF GVDPVTSQNV QYFLDRFYMS RISIRMLLNQ HSLLFGGKGK GSPSHRKHIG SINPNCNVLE VIKDGYENAR RLCDLYYINS PELELEELNA KSPGQPIQVV YVPSHLYHMVFELFKNAMRA TMEHHANRGV YPPIQVHVTL GNEDLTVKMS DRGGGVPLRK IDRLFNYMYS TAPRPRVETS RAVPLAGFGY GLPISRLYAQ YFQGDLKLYS LEGYGTDAVI YIKALSTDSI ERLPVYNKAA WKHYNTNHEA DDWCVPSREP KDMTTFRSA | |
Greater than 95.0% as determined by SDS-PAGE. |