missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDK1 Protein
A cDNA sequence encoding the PDK1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12998-50ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
PDK1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSSDSGSSPA SERGVPGQVD FYARFSPSPL SMKQFLDFGS VNACEKTSFM FLRQELPVRL ANIMKEISLLPDNLLRTPSV QLVQSWYIQS LQELLDFKDK SAEDAKAIYD FTDTVIRIRN RHNDVIPTMA QGVIEYKESF GVDPVTSQNV QYFLDRFYMS RISIRMLLNQ HSLLFGGKGK GSPSHRKHIG SINPNCNVLE VIKDGYENAR RLCDLYYINS PELELEELNA KSPGQPIQVV YVPSHLYHMVFELFKNAMRA TMEHHANRGV YPPIQVHVTL GNEDLTVKMS DRGGGVPLRK IDRLFNYMYS TAPRPRVETS RAVPLAGFGY GLPISRLYAQ YFQGDLKLYS LEGYGTDAVI YIKALSTDSI ERLPVYNKAA WKHYNTNHEA DDWCVPSREP KDMTTFRSA | |
Greater than 95.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PDK1 solution containing 20mM Tris pH 7, 0.5mM DTT, 0.1M NaCl, 0.1mM EDTA 0.1mM PMSF, 1mM MgCl2 and 40% glycerol. |