missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDE6D Protein
A cDNA sequence encoding the PDE6D was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 5210.00 CHF
Spezifikation
Gen-ID (Entrez) | 5147 |
---|---|
Name | PDE6D Protein |
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gensymbol | PDE6D |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15992169
|
enQuireBio™
QP12990-5ug |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15982169
|
enQuireBio™
QP12990-20ug |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15972169
|
enQuireBio™
QP12990-1mg |
1 mg |
5210.00 CHF
1 Milligramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Specifications
5147 | |
Research Use Only | |
PDE6D | |
Human | |
His | |
The PDE6D protein solution contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT and 100mM NaCl. |
PDE6D Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MSAKDERARE ILRGFKLNWM NLRDAETGKI LWQGTEDLSV PGVEHEARVP KKILKCKAVS RELNFSSTEQ MEKFRLEQKV YFKGQCLEEW FFEFGFVIPN STNTWQSLIE AAPESQMMPA SVLTGNVIIE TKFFDDDLLV STSRVRLFYV GSHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |