Learn More
enQuireBio™ Recombinant Human PDE6D Protein
A cDNA sequence encoding the PDE6D was constructed and used to recombinantly synthesize the protein.
Marke:  enQuireBio™ QP12990-5ug
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15992169
  • 141.00 CHF / 5 Mikrogramm
Verfügbar ab: 03-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Spezifikation
5147
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MSAKDERARE ILRGFKLNWM NLRDAETGKI LWQGTEDLSV PGVEHEARVP KKILKCKAVS RELNFSSTEQ MEKFRLEQKV YFKGQCLEEW FFEFGFVIPN STNTWQSLIE AAPESQMMPA SVLTGNVIIE TKFFDDDLLV STSRVRLFYV GSHHHHHH
Greater than 90.0% as determined by SDS-PAGE.
PDE6D Protein
Research Use Only
PDE6D
Human
His
The PDE6D protein solution contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT and 100mM NaCl.