missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human NSF GST (N-Term) Protein
Recombinant Protein
Marke: Novus Biologicals™ H00004905-P01-10ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Spezifikation
Western Blot, ELISA, Immunoprecipitation, Protein Array, Immunoaffinity Purification | |
4905 | |
10 μg | |
EC 3.6.4.6, NEM-sensitive fusion protein, N-ethylmaleimide-sensitive factor, N-ethylmaleimide-sensitive factor-like protein, N-ethylmaleimide-sensitive fusion protein, SKD2, vesicle-fusing ATPase, Vesicular-fusion protein NSF | |
Use in Immunoprecipitation reported in scientific literature (PMID 25972187) | |
NSF |
50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
MolecularWeight-theroretical: 109 kDa | |
Store at -80°C. Avoid freeze-thaw cycles. | |
NSF | |
>80% by SDS-PAGE and Coomassie blue staining |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Novus Biologicals™ Recombinant Human NSF GST (N-Term) Protein > 10 μg
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur