missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFAF1 Protein
A cDNA sequence encoding the NDUFAF1 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 5210.00 CHF
Spezifikation
Gen-ID (Entrez) | 51103 |
---|---|
Name | NDUFAF1 Protein |
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gensymbol | NDUFAF1 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15991629
|
enQuireBio™
QP12813-5ug |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15981629
|
enQuireBio™
QP12813-20ug |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15971629
|
enQuireBio™
QP12813-1mg |
1 mg |
5210.00 CHF
1 Milligramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
51103 | |
Research Use Only | |
NDUFAF1 | |
Human | |
His | |
The NDUFAF1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
NDUFAF1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAAAAS RGVGAKLGLR EIRIHLCQRS PGSQGVRDFI EKRYVELKKA NPDLPILIRE CSDVQPKLWA RYAFGQETNV PLNNFSADQV TRALENVLSG KA | |
Greater than 90.0% as determined by SDS-PAGE. |