missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human METAP1D Protein
A cDNA sequence encoding the METAP1D was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12685-1mg
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
METAP1D Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSSPLNHI YLHKQSSSQQ RRNFFFRRQR DISHSIVLPA AVSSAHPVPK HIKKPDYVTT GIVPDWGDSI EVKNEDQIQG LHQACQLARH VLLLAGKSLK VDMTTEEIDA LVHREIISHN AYPSPLGYGG FPKSVCTSVN NVLCHGIPDS RPLQDGDIIN IDVTVYYNGY HGDTSETFLV GNVDECGKKL VEVARRCRDE AIAACRAGAP FSVIGNTISH ITHQNGFQVC PHFVGHGIGS YFHGHPEIWH HANDSDLPME EGMAFTIEPI ITEGSPEFKV LEDAWTVVSL DNQRSAQFEH TVLITSRGAQ ILTKLPHEA | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The METAP1D solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 20% glycerol and 1mM DTT. |