Learn More
R&D Systems™ Recombinant Human IL-7 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke: R&D Systems™ 207-IL-005/CF
132.80 CHF gültig bis 2025-07-30
Benutzen Sie den Promotions-Code "24078" um Ihren Promotions-Preis zu erhalten.
Beschreibung
Measured in a cell proliferation assay using PHA-activated human peripheral blood lymphocytes (PBL). Yokota, T. et al. (1986) Proc. Natl. Acad. Sci. USA 83:5894. The ED50 for this effect is 0.1-0.5 ng/mL. The specific activity of Recombinant Human IL-7 is approximately 4.4 x 105 IU/μg, which is calibrated against human IL-7 WHO International Standard (NIBSC code: 90/530). Specific activity is for reference purposes only and is not routinely tested.
Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS. | |
17 kDa | |
5 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
Reconstitute 5 μg vials at 50 μg/mL in sterile PBS. Reconstitute 10 μg or larger vials (at 100 μg/mL in sterile PBS.) | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
3574 | |
IL-7 | |
E. coli-derived human IL-7 protein Asp26-His177,with an N-terminal Met | |
<0.01 EU per 1μg of the protein by the LAL method. | |
Recombinant | |
Lyophilized from a 0.2μm filtered solution in PBS. |
Aktionen
-
R&D Systems
,
Tocris
Ultimative Promo Mai
Forschung fördern: Die richtigen Reagenzien für jeden SchrittAktionscode: 24078Gültig bis zum: 2025-07-30
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.