missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HFE Protein
A cDNA sequence encoding the HFE was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12227-1mg
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
HFE Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMRLLRSH SLHYLFMGAS EQDLGLSLFE ALGYVDDQLF VFYDHESRRV EPRTPWVSSR ISSQMWLQLS QSLKGWDHMF TVDFWTIMEN HNHSKESHTL QVILGCEMQE DNSTEGYWKY GYDGQDHLEF CPDTLDWRAA EPRAWPTKLE WERHKIRARQ NRAYLERDCP AQLQQLLELG RGVLDQQVPP LVKVTHHVTS SVTTLRCRAL NYYPQNITMK WLKDKQPMDA KEFEPKDVLP NGDGTYQGWI TLAVPPGEEQ RYTCQVEHPG LDQPLIVIWE PSPSGTLV | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HFE protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |