Learn More
enQuireBio™ Recombinant Human GNAI1 Protein
A cDNA sequence encoding the GNAI1 was constructed and used to recombinantly synthesize the protein.
141.00 CHF - 8085.00 CHF
Spezifikation
Name | GNAI1 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15979078
|
enQuireBio™
QP12009-2ug |
2 μg |
141.00 CHF
2 Mikrogramm |
Verfügbar ab: 06-03-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15959078
|
enQuireBio™
QP12009-10ug |
10 μg | N/A | ||||||
15969078
|
enQuireBio™
QP12009-1mg |
1 mg |
8085.00 CHF
1 Milligramm |
Verfügbar ab: 06-03-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
GNAI1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GNAI1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGCTLSA EDKAAVERSK MIDRNLREDG EKAAREVKLL LLGAGESGKS TIVKQMKIIH EAGYSEEECK QYKAVVYSNT IQSIIAIIRA MGRLKIDFGD SARADDARQL FVLAGAAEEG FMTAELAGVI KRLWKDSGVQ ACFNRSREYQ LNDSAAYYLN DLDRIAQPNY IPTQQDVLRT RVKTTGIVET HFTFKDLHFK MFDVGGQRSE RKKWIHCFEG VTAIIFCVAL SDYDLVLAED EEMNRMHESM KLFDSICNNK WFTDTSIILF LNKKDLFEEK IKKSPLTICY PEYAGSNTYE EAAAYIQCQF EDLNKRKDTK EIYTHFTCAT DTKNVQFVFD AVTDVIIKNN LKDCGLF | |
Greater than 90.0% as determined by SDS-PAGE. |