missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Egl nine homolog 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Egl nine homolog 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5968-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_444274.1 | |
Egl nine homolog 2 Protein | |
Research Use Only | |
EGLN2 | |
Recombinant Protein | |
E. coli | |
MVACYPGNGL GYVRHVDNPH GDGRCITCIY YLNQNWDVKV HGGLLQIFPE GRPVVANIEP LFDRLLIFWS DRRNPHEVKP AYATRYAITV WYFDAKERAA AKDKYQLASG QKGVQVPVSQ PPTPT | |
Greater than 90% as determined by SDS-PAGE. |
112398 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction