missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Cyclophilin A Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Marke: R&D Systems™ 3589-CAB-100
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Recombinant Human Cyclophilin A Protein is derived from E. coli. The Recombinant Human Cyclophilin A Protein has been validated for the following applications: Enzyme Activity.
Spezifikation
P62937 | |
5478 | |
100 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening. | |
<1.0 EU per 1 μg of the protein by the LAL method. | |
Unconjugated | |
Recombinant | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining. |
Enzyme Activity | |
18 kDa | |
E. coli-derived human Cyclophilin A protein Met1-Glu165 | |
RUO | |
Cyclophilin A | |
Human | |
Supplied as a 0.2 μm filtered solution in MES and NaCl. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
R&D Systems™ Recombinant Human Cyclophilin A Protein, CF > Quantity: 100 μg
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur