Learn More
R&D Systems™ Recombinant Human CCL23/MPIF-1 (aa 46-120) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke: R&D Systems™ 131-M1-025
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Recombinant Human CCL23/MPIF-1 (aa 46-120) Protein is derived from E. coli. The Recombinant Human CCL23/MPIF-1 (aa 46-120) Protein has been validated for the following applications: Bioactivity.Spezifikation
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | |
8.6 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
6368 | |
CCL23/MPIF-1 aa 46-120 | |
E. coli-derived human CCL23/MPIF-1 protein Arg46-Asn120 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |