Learn More
R&D Systems™ Recombinant Human CCL23/MPIF-1 (aa 46-120) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke:  R&D Systems™ 131-M1-025
 Lernen Sie weitere Varianten dieses
Artikelnummer. 18177630
  • 349.00 CHF / 25 Mikrogramm
Verfügbar ab: 05-02-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The Recombinant Human CCL23/MPIF-1 (aa 46-120) Protein is derived from E. coli. The Recombinant Human CCL23/MPIF-1 (aa 46-120) Protein has been validated for the following applications: Bioactivity.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
8.6 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
6368
CCL23/MPIF-1 aa 46-120
E. coli-derived human CCL23/MPIF-1 protein Arg46-Asn120
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.