missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 Protein
A cDNA sequence encoding the HIV-1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12240-500ug
This item is not returnable.
View return policy
Specifications
HIV-1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
100mM NaPO4, pH 6 and 0.05% SDS. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
Greater than 95.0% as determined by HPLC analysis and SDS-PAGE. |