missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis A HAV P3C Protein
A cDNA sequence encoding the HAV P3C was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12110-1mg
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
Hepatitis A HAV P3C Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
10mM CBB, pH9.6, 0.1% SDS and 50% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis A | |
Untagged | |
HAV P3C protein is >90% pure as determined by 10% PAGE(coomassie staining). |