missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Goat IgG Recombinant Secondary Antibody, Alexa Fluor™ Plus 647, Invitrogen™

Rabbit Recombinant Monoclonal Secondary Antibody

107.00 CHF - 260.00 CHF

Spezifikation

Antigen Goat IgG
Klon 21H44L11
Konzentration 1 mg/mL
Anwendungen Flow Cytometry, Immunocytochemistry, Western Blot
Klassifikation Recombinant Monoclonal
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
17855672
Unterlagen ansehen
Invitrogen
A5657050UL
50 μL
107.00 CHF
50 Mikroliter
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
17846562
Unterlagen ansehen Aktionsdetails
Invitrogen
A56570
250 μL
260.00 CHF
250 Mikroliter
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity and superior lot-to-lot consistency. This recombinant antibody has been engineered with point mutations that eliminate binding to Fc receptors, enabling reduction in background and superior signal to noise. Obtain superior images with the new Invitrogen Alexa Fluor Plus secondary antibodies. Make your low abundant targets visible, minimize time spent optimizing, and make your precious samples count. Alexa Fluor Plus secondary antibodies provide brighter signal compared to leading Alexa Fluor secondary antibodies, providing you with better signal-to-noise for your critical experiments. Alexa Fluor Plus secondary antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor Plus secondary antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning images. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically.

Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation

Spezifikation

Goat IgG
1 mg/mL
Recombinant Monoclonal
Liquid
RUO
proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.5
Secondary
4° C, store in dark, DO NOT FREEZE!
21H44L11
Flow Cytometry, Immunocytochemistry, Western Blot
Alexa Fluor Plus 647
Rabbit
Goat
IgG
Affinity chromatography
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Rabbit anti-Goat IgG Recombinant Secondary Antibody, Alexa Fluor™ Plus 647, Invitrogen™

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt