Learn More
R&D Systems™ TACS XTT Cell Proliferation/Viability Assay
Avoids radioactivity and gives reproducible and sensitive results
Marke: R&D Systems™ 4891-025-K
Includes: XTT Reagent, 5 x 25mL; XTT Activator, 5 x 0.5mL
Beschreibung
XTT is added directly to the culture medium and cleavage of the tetrazolium salt to formazan occurs in metabolically active cells
The orange formazan dye generated can be measured by absorbance at 490nm (or 450nm) in a microplate reader to yield rapid re
Spezifikation
2500 Tests | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |