Learn More
R&D Systems™ Human KIR3DL3/CD158z (AAC99763) VersaClone cDNA
Killer Cell Immunoglobulin-like Receptor, Three Domain Long Cytoplasmic Tail, 3
Marke: R&D Systems™ RDC0575
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Human KIR3DL3/CD158z (AAC99763) VersaClone cDNA clone contains the complete ORF for KIR3DL3/CD158z, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The KIR3DL3/CD158z insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Bestellinformation
Ships at ambient temperature.
Spezifikation
hKIR3DL3 | |
10 μg at 0.2 μg/μL | |
1246bp | |
Human |
AAC99763 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
Berichtigung von Produktinhalten
Name des Produkts
R&D Systems™ Human KIR3DL3/CD158z (AAC99763) VersaClone cDNA > 10 μg
For Reasearch Use Only; Not for Use in Humans
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur