Learn More
POMC Mouse anti-Human, CoraLite 594, Clone: 3C12C7, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL594-66358-100UL
Beschreibung
The POMC antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human POMC. This antibody recognizes human, rat, mouse antigen. The POMC antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Spezifikation
POMC | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat |
Immunofluorescence | |
3C12C7 | |
CoraLite 594 | |
100 μL | |
5443 | |
IgG1 |